missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4,6-dichloropyrimidine, 95%, tech., Thermo Scientific Chemicals

Catalog Number p-3777251
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
56-05-3
C4H3Cl2N3
163.99
MFCD00006090
JPZOAVGMSDSWSW-UHFFFAOYSA-N
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351
65522
4,6-dichloropyrimidin-2-amine
C1=C(N=C(N=C1Cl)N)Cl
This item is not returnable. View return policy
56-05-3
C4H3Cl2N3
163.99
MFCD00006090
JPZOAVGMSDSWSW-UHFFFAOYSA-N
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351
65522
4,6-dichloropyrimidin-2-amine
C1=C(N=C(N=C1Cl)N)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 219°C to 226°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 10 g
Beilstein 24, 81
Formula Weight 163.99
Percent Purity 95%
Grade Technical
Physical Form Liquid
Chemical Name or Material 2-Amino-4, 6-dichloropyrimidine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-4,6-dichloropyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'