missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-4-(2-hydroxyphenyl)thiazole, 97%, Thermo Scientific™

Catalog Number 15464448
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
78546-66-4
C11H10N2O2S
234.273
MFCD00665950
RCVLPHKAZAQDGZ-UHFFFAOYSA-N
2-acetamido-4-2-hydroxyphenyl thiazole, n-4-2-hydroxyphenyl-1,3-thiazol-2-yl acetamide, 2-acetylamino-4-2-hydroxyphenyl thiazole, n-4-2-hydroxyphenyl thiazol-2-yl acetamide
73996299
N-[4-(2-hydroxyphenyl)-1,3-thiazol-2-yl]acetamide
CC(=O)NC1=NC(=CS1)C2=CC=CC=C2O
This item is not returnable. View return policy
78546-66-4
C11H10N2O2S
234.273
MFCD00665950
RCVLPHKAZAQDGZ-UHFFFAOYSA-N
2-acetamido-4-2-hydroxyphenyl thiazole, n-4-2-hydroxyphenyl-1,3-thiazol-2-yl acetamide, 2-acetylamino-4-2-hydroxyphenyl thiazole, n-4-2-hydroxyphenyl thiazol-2-yl acetamide
73996299
N-[4-(2-hydroxyphenyl)-1,3-thiazol-2-yl]acetamide
CC(=O)NC1=NC(=CS1)C2=CC=CC=C2O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 224°C to 228°C
Quantity 2 g
Beilstein 18070656
Formula Weight 234.27
Percent Purity 97%
Chemical Name or Material 2-Acetamido-4-(2-hydroxyphenyl)thiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'