missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-4-methoxyaniline, 97%, Thermo Scientific™

Catalog Number p-7029281
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
151414-47-0
C7H7F2NO
159.136
MFCD04115922
HQBURSNJKAITGQ-UHFFFAOYSA-N
4-amino-3,5-difluoroanisole, 2,6-difluoro-4-methoxyphenylamine, benzenamine, 2,6-difluoro-4-methoxy, 2,6-difluoro-4-methoxy-benzenamine, 2,6-difluoro-4-methoxy-phenylamine, intermediates-zcf02157, acmc-209z8h, 4-methoxy-2,6-difluoroaniline, 2,6-difluoro-4-methoxybenzenamine, benzenamine,2,6-difluoro-4-methoxy
5185763
2,6-difluoro-4-methoxyaniline
COC1=CC(=C(C(=C1)F)N)F
This item is not returnable. View return policy
151414-47-0
C7H7F2NO
159.136
MFCD04115922
HQBURSNJKAITGQ-UHFFFAOYSA-N
4-amino-3,5-difluoroanisole, 2,6-difluoro-4-methoxyphenylamine, benzenamine, 2,6-difluoro-4-methoxy, 2,6-difluoro-4-methoxy-benzenamine, 2,6-difluoro-4-methoxy-phenylamine, intermediates-zcf02157, acmc-209z8h, 4-methoxy-2,6-difluoroaniline, 2,6-difluoro-4-methoxybenzenamine, benzenamine,2,6-difluoro-4-methoxy
5185763
2,6-difluoro-4-methoxyaniline
COC1=CC(=C(C(=C1)F)N)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluoro-4-methoxyaniline is used as primary and secondary intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 38°C to 40°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 159.14
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-4-methoxyaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'