missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluoro-3-methylaniline, 97%, Thermo Scientific™

Catalog Number p-7029216
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
144851-63-8
C7H7F2N
143.14
MFCD06660182
ZMNJSSZHDRBGNN-UHFFFAOYSA-N
benzenamine, 2,6-difluoro-3-methyl, 2,6-difluoro-m-toluidine, 3-amino-2,4-difluorotoluene, 2,6-difluoro-3-methylphenylamine
15017536
2,6-difluoro-3-methylaniline
CC1=C(F)C(N)=C(F)C=C1
This item is not returnable. View return policy
144851-63-8
C7H7F2N
143.14
MFCD06660182
ZMNJSSZHDRBGNN-UHFFFAOYSA-N
benzenamine, 2,6-difluoro-3-methyl, 2,6-difluoro-m-toluidine, 3-amino-2,4-difluorotoluene, 2,6-difluoro-3-methylphenylamine
15017536
2,6-difluoro-3-methylaniline
CC1=C(F)C(N)=C(F)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,6-Difluoro-3-methylaniline is used as a solvent is convenient as the pyridine can serve both as the solvent and the catalyst in the transformation.

Solubility
Soluble in organic solvents.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2810
Solubility Information Soluble in organic solvents.
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-3-methylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'