missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloropyridine-3-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7030069
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1073371-78-4
C11H14BCl2NO2
273.948
MFCD06798261
AOLXLWZJOQBPQX-UHFFFAOYSA-N
2,6-dichloropyridine-3-boronic acid pinacol ester, 2,6-dichloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dichloropyridin-3-ylboronic acid pinacol ester, pyridine, 2,6-dichloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5532, 2,6-dichloropyridine-3-boronicacidpinacolester, 2,6-dichloropyridin-3-yl boronic acid pinacol ester, 2,6-dichloro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
44755160
2,6-dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)Cl)Cl
This item is not returnable. View return policy
1073371-78-4
C11H14BCl2NO2
273.948
MFCD06798261
AOLXLWZJOQBPQX-UHFFFAOYSA-N
2,6-dichloropyridine-3-boronic acid pinacol ester, 2,6-dichloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dichloropyridin-3-ylboronic acid pinacol ester, pyridine, 2,6-dichloro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5532, 2,6-dichloropyridine-3-boronicacidpinacolester, 2,6-dichloropyridin-3-yl boronic acid pinacol ester, 2,6-dichloro-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
44755160
2,6-dichloro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=C(C=C2)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 51°C to 53°C
Quantity 250 mg
Formula Weight 273.96
Percent Purity 95%
Chemical Name or Material 2,6-Dichloropyridine-3-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'