missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-7031025
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
25922-41-2
C8H3Cl2F3O2
259.01
MFCD09743917
WRNSEKNHEIXBES-UHFFFAOYSA-N
2,6-dichloro-3-trifluoromethyl benzoic acid, 2,6-dichloro-3-trifluoromethyl-benzoic acid, 2,6-dichloro-3-trifluoromethyl benzoic acid, jrd
11680459
2,6-dichloro-3-(trifluoromethyl)benzoic acid
OC(=O)C1=C(Cl)C=CC(=C1Cl)C(F)(F)F
This item is not returnable. View return policy
25922-41-2
C8H3Cl2F3O2
259.01
MFCD09743917
WRNSEKNHEIXBES-UHFFFAOYSA-N
2,6-dichloro-3-trifluoromethyl benzoic acid, 2,6-dichloro-3-trifluoromethyl-benzoic acid, 2,6-dichloro-3-trifluoromethyl benzoic acid, jrd
11680459
2,6-dichloro-3-(trifluoromethyl)benzoic acid
OC(=O)C1=C(Cl)C=CC(=C1Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 102°C
Quantity 1 g
Formula Weight 259.01
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'