missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-nitropyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7025645
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16013-85-7
C5H2Cl2N2O2
192.983
MFCD00006234
SHCWQWRTKPNTEM-UHFFFAOYSA-N
2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n
85239
2,6-dichloro-3-nitropyridine
C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16013-85-7
C5H2Cl2N2O2
192.983
MFCD00006234
SHCWQWRTKPNTEM-UHFFFAOYSA-N
2,6-dichloro-3-nitro-pyridine, pyridine, 2,6-dichloro-3-nitro, 3-nitro-2,6-dichloropyridine, 2,6-dichloro-3-nitro pyridine, zlchem 223, pubchem9234, acmc-209dko, 2,6-dichloronitropyridine, dichloronitropyridine, ksc174q3n
85239
2,6-dichloro-3-nitropyridine
C1=CC(=NC(=C1[N+](=O)[O-])Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 62°C to 65°C
Flash Point >110°C (230°F)
Quantity 10 g
Formula Weight 192.99
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'