missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-iodopyridine, 98%, Thermo Scientific™

Catalog Number p-7055717
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
148493-37-2
C5H2Cl2IN
273.882
MFCD03094686
DPCQIHCGMIPSQV-UHFFFAOYSA-N
2736002
2,6-dichloro-3-iodopyridine
C1=CC(=NC(=C1I)Cl)Cl
This item is not returnable. View return policy
148493-37-2
C5H2Cl2IN
273.882
MFCD03094686
DPCQIHCGMIPSQV-UHFFFAOYSA-N
2736002
2,6-dichloro-3-iodopyridine
C1=CC(=NC(=C1I)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 74°C to 79°C
Color White to Brown
Quantity 1 g
UN Number UN2811
Beilstein 6135696
Sensitivity Light sensitive
Synonym 2,6-dichloro-3-iodo-pyridine, pyridine, 2,6-dichloro-3-iodo, zlchem 993, acmc-20aihk, pubchem14239, tpc-py080, 2,6-dichloro-3-iodo pyridine, pyridine,2,6-dichloro-3-iodo, 2,6-dichloro-3-iodopyridine
Solubility Information Slightly soluble in water.
Formula Weight 273.88
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,6-Dichloro-3-iodopyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'