missing translation for 'onlineSavingsMsg'
Learn More

2,5-dimethylphenyl isothiocyanate, Thermo Scientific™

Catalog Number p-4391329
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
19241-15-7
C9H9NS
163.238
RCBXZRJVMSWGAO-UHFFFAOYSA-N
140499
2-isothiocyanato-1,4-dimethylbenzene
CC1=CC(=C(C=C1)C)N=C=S
This item is not returnable. View return policy
19241-15-7
C9H9NS
163.238
RCBXZRJVMSWGAO-UHFFFAOYSA-N
140499
2-isothiocyanato-1,4-dimethylbenzene
CC1=CC(=C(C=C1)C)N=C=S
Assay Percent Range 97%
Synonym 2,5-dimethylphenyl isothiocyanate, benzene,2-isothiocyanato-1,4-dimethyl, 2,5-dimethylbenzenisothiocyanate, 2-isothiocyanato-1,4-dimethyl-benzene, 2,5-xylyl isothiocyanate, 2,5-dimethylphenyl-isothiocyanate, 2,5-dimethylphenylisothiocyanate, #, benzene, 2-isothiocyanato-1,4-dimethyl
Formula Weight 163.24
Percent Purity 97%
Quantity 5 g
Packaging Amber glass bottle
Chemical Name or Material 2, 5-Dimethylphenyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'