Learn More
2,5-Difluorobenzenesulfonyl chloride, 97%, Thermo Scientific™
Catalog Number p-4089896
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
26120-86-5
C6H3ClF2O2S
212.3
MFCD00060682
CELLJWUVMKEJDY-UHFFFAOYSA-N
2,5-difluorobenzene-1-sulfonyl chloride, 2,5-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,5-difluoro, 2,5-difluoro-benzenesulfonyl chloride, 2,5-difluorobenzenesulphonylchloride, pubchem5123, acmc-1ch2j, ksc201g8t, zerenex zx009573
2736911
2,5-difluorobenzenesulfonyl chloride
C1=CC(=C(C=C1F)S(=O)(=O)Cl)F
26120-86-5
C6H3ClF2O2S
212.3
MFCD00060682
CELLJWUVMKEJDY-UHFFFAOYSA-N
2,5-difluorobenzene-1-sulfonyl chloride, 2,5-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,5-difluoro, 2,5-difluoro-benzenesulfonyl chloride, 2,5-difluorobenzenesulphonylchloride, pubchem5123, acmc-1ch2j, ksc201g8t, zerenex zx009573
2736911
2,5-difluorobenzenesulfonyl chloride
C1=CC(=C(C=C1F)S(=O)(=O)Cl)F
CAS Min % 96.0
CAS Max % 100.0
Density 1.5800g/mL
Boiling Point 221°C
Flash Point 110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (F)2C6H3SO2Cl
Quantity 1 g
Specific Gravity 1.58
Formula Weight 212.3
Percent Purity 97%
Chemical Name or Material 2, 5-Difluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Difluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.