missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloropyridine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
50 g
250 g
Description
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.
Solubility
Insoluble in water.
Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
2,5-Dichloropyridine was used in the synthesis of 6-halo-pyridin-3-yl boronic acids and esters. It undergoes cross-coupling reaction with arylboronic acids in the presence of [1,4-bis-(diphenylphosphine)butane]palladium (II) dichloride as catalyst.
Solubility
Insoluble in water.
Notes
Incompatible with Strong acids, strong bases, strong oxidizing agents.
Specifications
Melting Point | 59°C to 62°C |
Boiling Point | 190°C to 191°C |
Flash Point | 112°C (233°F) |
Odor | Unpleasant |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 108886 |
Solubility Information | Insoluble in water. |
Formula Weight | 147.99 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'