missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7030153
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1073353-98-6
C11H14BCl2NO2
273.948
MFCD06798256
ROOZGFDFEKVFDG-UHFFFAOYSA-N
2,5-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,5-dichloropyridine-4-boronic acid pinacol ester, pyridine, 2,5-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5530, 2,5-dichloropyridine-4-boronic acid, pinacol ester, 2,5-dichloropyridin-4-yl boronic acid pinacol ester, 2,5-dichloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
44755157
2,5-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2Cl)Cl
This item is not returnable. View return policy
1073353-98-6
C11H14BCl2NO2
273.948
MFCD06798256
ROOZGFDFEKVFDG-UHFFFAOYSA-N
2,5-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,5-dichloropyridine-4-boronic acid pinacol ester, pyridine, 2,5-dichloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-13-5530, 2,5-dichloropyridine-4-boronic acid, pinacol ester, 2,5-dichloropyridin-4-yl boronic acid pinacol ester, 2,5-dichloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
44755157
2,5-dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=CC(=NC=C2Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 83°C to 85°C
Quantity 250 mg
Formula Weight 273.96
Percent Purity 95%
Chemical Name or Material 2,5-Dichloropyridine-4-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'