missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichlorobenzylmagnesium chloride, 0.25M in 2-MeTHF, Thermo Scientific™
Click to view available options
Quantity:
100 mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 213627-19-1 |
Molecular Formula | C7H5Cl3Mg |
MDL Number | MFCD06200641 |
Quantity | 100 mL |
UN Number | UN2924 |
Sensitivity | Air and moisture sensitive |
Synonym | 2,5-dichlorobenzylmagnesium chloride, 2,5-dichlorobenzylmagnesium chloride, 0.25m in 2-methf, chloro 2,5-dichlorophenyl methyl magnesium, gcykxkkuuljojj-uhfffaoysa-m, 2,5-dichlorobenzyl magnesium chloride, magnesium,1,4-dichloro-2-methanidylbenzene,chloride, 2,5-dichlorobenzylmagnesium chloride 0.25 m in diethyl ether |
InChI Key | LCSTXMZCRKYVRE-UHFFFAOYSA-M |
SMILES | [Mg++].[Cl-].[CH2-]C1=C(Cl)C=CC(Cl)=C1 |
IUPAC Name | magnesium;1,4-dichloro-2-methanidylbenzene;chloride |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'