Learn More
2,5-Dichlorobenzyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
Specifications
Boiling Point | 121°C to 124°C (14 mmHg) |
Odor | Pungent |
Refractive Index | 1.5755 |
Quantity | 5 g |
UN Number | UN3265 |
Beilstein | 637802 |
Solubility Information | Not miscible in water. |
Formula Weight | 195.48 |
Percent Purity | 97% |
Chemical Name or Material | 2,5-Dichlorobenzyl chloride |