Learn More
2,4-dimethyl-3-furaldehyde, Thermo Scientific™
Catalog Number p-4391323
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
75002-34-5
C7H8O2
124.139
MGSAYHDYMMHZQE-UHFFFAOYSA-N
2,4-dimethyl-3-furaldehyde, 3-furancarbaldehyde,2,4-dimethyl, 3-furancarboxaldehyde,2,4-dimethyl, 3-furancarboxaldehyde, 2,4-dimethyl-9ci
21702758
2,4-dimethylfuran-3-carbaldehyde
CC1=COC(=C1C=O)C
75002-34-5
C7H8O2
124.139
MGSAYHDYMMHZQE-UHFFFAOYSA-N
2,4-dimethyl-3-furaldehyde, 3-furancarbaldehyde,2,4-dimethyl, 3-furancarboxaldehyde,2,4-dimethyl, 3-furancarboxaldehyde, 2,4-dimethyl-9ci
21702758
2,4-dimethylfuran-3-carbaldehyde
CC1=COC(=C1C=O)C
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 124.14
Percent Purity 95%
Chemical Name or Material 2, 4-dimethyl-3-furaldehyde