missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-4510588
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
95-01-2
C7H6O3
138.12
MFCD00011686
IUNJCFABHJZSKB-UHFFFAOYSA-N
beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehyde
7213
CHEBI:50198
2,4-dihydroxybenzaldehyde
OC1=CC=C(C=O)C(O)=C1
95-01-2
C7H6O3
138.12
MFCD00011686
IUNJCFABHJZSKB-UHFFFAOYSA-N
beta-resorcylaldehyde, 4-hydroxysalicylaldehyde, 4-formylresorcinol, beta-resorcaldehyde, benzaldehyde, 2,4-dihydroxy, beta-resorcinaldehyde, beta-resorcylic aldehyde, 2,4-dihydroxybenzenecarbonal, salicylaldehyde, 4-hydroxy, 4-hydroxysalicyladehyde
7213
CHEBI:50198
2,4-dihydroxybenzaldehyde
OC1=CC=C(C=O)C(O)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 133.0°C to 138.0°C
Color White to Beige
Boiling Point 220.0°C to 228.0°C (22.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (GC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 25 g
Beilstein 08,241
Merck Index 15,8277
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in alcohol,ether,chloroform and,glacial acetic acic
Formula Weight 138.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dihydroxybenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'