missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorophenyl isothiocyanate, 96%, Thermo Scientific Chemicals

Catalog Number p-7024934
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
141106-52-7
C7H3F2NS
171.165
MFCD00041044
ABGGPKIFVAIRGU-UHFFFAOYSA-N
2,4-difluorophenyl isothiocyanate, 2,4-difluoro isothiocyanato benzene, 2,4-difluoro-1-isothiocyanato-benzene, 2,4-difluorophenylisothiocyanate, isothiocyanic acid 2,4-difluorophenyl ester, benzene, 2,4-difluoro-1-isothiocyanato, benzene, 2,4-difluoro-1-isothiocyanato-9ci, 2,4-difluorobenzenisothiocyanate, pubchem4451, acmc-209clq
518879
2,4-difluoro-1-isothiocyanatobenzene
C1=CC(=C(C=C1F)F)N=C=S
This item is not returnable. View return policy
141106-52-7
C7H3F2NS
171.165
MFCD00041044
ABGGPKIFVAIRGU-UHFFFAOYSA-N
2,4-difluorophenyl isothiocyanate, 2,4-difluoro isothiocyanato benzene, 2,4-difluoro-1-isothiocyanato-benzene, 2,4-difluorophenylisothiocyanate, isothiocyanic acid 2,4-difluorophenyl ester, benzene, 2,4-difluoro-1-isothiocyanato, benzene, 2,4-difluoro-1-isothiocyanato-9ci, 2,4-difluorobenzenisothiocyanate, pubchem4451, acmc-209clq
518879
2,4-difluoro-1-isothiocyanatobenzene
C1=CC(=C(C=C1F)F)N=C=S

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.349
Boiling Point 98°C to 99°C (30 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.597
Quantity 1 g
UN Number UN2922
Beilstein 6919744
Sensitivity Moisture sensitive
Formula Weight 171.17
Percent Purity 96%
Chemical Name or Material 2,4-Difluorophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'