missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-1-fluorobenzene, 98%, Thermo Scientific™

Catalog Number p-7048059
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1435-53-6
C6H3Br2F
253.90
MFCD00000283
MKHDDTWHDFVYDQ-UHFFFAOYSA-N
2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j
15034
2,4-dibromo-1-fluorobenzene
FC1=CC=C(Br)C=C1Br
This item is not returnable. View return policy
1435-53-6
C6H3Br2F
253.90
MFCD00000283
MKHDDTWHDFVYDQ-UHFFFAOYSA-N
2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j
15034
2,4-dibromo-1-fluorobenzene
FC1=CC=C(Br)C=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 2.047
Boiling Point 103°C (32 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.584
Quantity 5 g
Beilstein 1934822
Formula Weight 253.91
Percent Purity 98%
Chemical Name or Material 2,4-Dibromo-1-fluorobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'