missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenyl)pyrrolidine, 95%, Thermo Scientific™

Catalog Number p-7054010
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
38944-14-8
C10H12ClN
181.66
MFCD01631838
CIHHGGKKRPPWSU-UHFFFAOYNA-N
2-4-chlorophenyl pyrrolidine, 2-4-chloro-phenyl-pyrrolidine, pyrrolidine, 2-4-chlorophenyl, 2-p-chlorophenyl pyrolidine, 2-p-chlorophenyl pyrrolidine, 2-4-chlorophenyl pyrrolidine #
592391
2-(4-chlorophenyl)pyrrolidine
ClC1=CC=C(C=C1)C1CCCN1
This item is not returnable. View return policy
38944-14-8
C10H12ClN
181.66
MFCD01631838
CIHHGGKKRPPWSU-UHFFFAOYNA-N
2-4-chlorophenyl pyrrolidine, 2-4-chloro-phenyl-pyrrolidine, pyrrolidine, 2-4-chlorophenyl, 2-p-chlorophenyl pyrolidine, 2-p-chlorophenyl pyrrolidine, 2-4-chlorophenyl pyrrolidine #
592391
2-(4-chlorophenyl)pyrrolidine
ClC1=CC=C(C=C1)C1CCCN1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 181.66
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(4-Chlorophenyl)pyrrolidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'