Learn More
2,4,6-Trimethoxybenzylamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog Number p-4228940
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
146548-59-6
C10H15NO3·ClH
233.7
BLFRMOOGAICNSZ-UHFFFAOYSA-N
2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine hydrochloride, pubchem7389, 2, 4, 6-trimethoxybenzylamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1
16211914
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride
COC1=CC(=C(C(=C1)OC)CN)OC.Cl
146548-59-6
C10H15NO3·ClH
233.7
BLFRMOOGAICNSZ-UHFFFAOYSA-N
2,4,6-trimethoxybenzylamine hydrochloride, 2,4,6-trimethoxyphenyl methanamine hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine hydrochloride, pubchem7389, 2, 4, 6-trimethoxybenzylamine hydrochloride, acmc-1c5nr, benzenemethanamine, 2,4,6-trimethoxy-, hydrochloride, 1-2,4,6-trimethoxyphenyl methanamine-hydrogen chloride 1/1
16211914
(2,4,6-trimethoxyphenyl)methanamine;hydrochloride
COC1=CC(=C(C(=C1)OC)CN)OC.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 207°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 233.7
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 4, 6-Trimethoxybenzylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2,4,6-Trimethoxybenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.