missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4',6'-Trihydroxyacetophenone hydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2',4',6'-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′,4′,6′-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
2′,4′,6′-Trihydroxyacetophenone hydrate is a compound that enhances bile secretion.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 218°C to 223°C |
Quantity | 5 g |
Beilstein | 1911197 |
Solubility Information | Soluble in water. |
Formula Weight | 168.15 (Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | 2',4',6'-Trihydroxyacetophenone hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'