missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tribromo-3-hydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-7044899
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
2737-22-6
C7H3Br3O2
358.81
MFCD00017320
FAWOIFAFFUDNJX-UHFFFAOYSA-N
3-hydroxy-2,4,6-tribromobenzaldehyde, benzaldehyde, 3-hydroxy-2,4,6-tribromo, benzaldehyde,2,4,6-tribromo-3-hydroxy, acmc-1cj0g, 3-08-00-00209 beilstein handbook reference, 2,4,6-tris bromanyl-3-oxidanyl-benzaldehyde
200803
2,4,6-tribromo-3-hydroxybenzaldehyde
OC1=C(Br)C=C(Br)C(C=O)=C1Br
This item is not returnable. View return policy
2737-22-6
C7H3Br3O2
358.81
MFCD00017320
FAWOIFAFFUDNJX-UHFFFAOYSA-N
3-hydroxy-2,4,6-tribromobenzaldehyde, benzaldehyde, 3-hydroxy-2,4,6-tribromo, benzaldehyde,2,4,6-tribromo-3-hydroxy, acmc-1cj0g, 3-08-00-00209 beilstein handbook reference, 2,4,6-tris bromanyl-3-oxidanyl-benzaldehyde
200803
2,4,6-tribromo-3-hydroxybenzaldehyde
OC1=C(Br)C=C(Br)C(C=O)=C1Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 119°C to 122°C
Quantity 10 g
Beilstein 2098037
Sensitivity Air Sensitive
Formula Weight 358.81
Percent Purity 98%
Chemical Name or Material 2,4,6-Tribromo-3-hydroxybenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'