missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 97%, Thermo Scientific Chemicals

Catalog Number p-7046872
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
2554-06-5
MFCD00040293
This item is not returnable. View return policy
2554-06-5
MFCD00040293

2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane is used as an intermediate in organic synthesis. It is used in the preparation of 4-vinyl-benzoic acid ethyl ester. Further, it is used to prepare styrene derivatives by cross-coupling with aryl bromide in the presence of palladium catalyst.

Solubility
Miscible with most organic solvents.

Notes
Moisture sensitive. Incompatible with acids, bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 110°C (10mmHg)
Melting Point -43°C
Quantity 50 g
Molecular Formula C12H24O4Si4
Beilstein 1807272
Flash Point 112°C (233°F)
Synonym 2,4,6,8-tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane, 2,4,6,8-tetramethyl-2,4,6,8-tetravinyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, cyclotetrasiloxane, 2,4,6,8-tetraethenyl-2,4,6,8-tetramethyl, 1,3,5,7-tetravinyl-1,3,5,7-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethyl-1,3,5,7-tetravinylcyclotetrasiloxane, 2,4,6,8-tetravinyl-2,4,6,8-tetramethylcyclotetrasiloxane, tetravinyltetramethylcyclotetrasiloxane, s-tetramethyltetravinylcyclotetrasiloxane, unii-3nt606t1vk, cyclotetrasiloxane, tetraethenyltetramethyl
Solubility Information Miscible with most organic solvents.
InChI Key VMAWODUEPLAHOE-UHFFFAOYSA-N
SMILES C[Si]1(O[Si](O[Si](O[Si](O1)(C)C=C)(C)C=C)(C)C=C)C=C
IUPAC Name 2,4,6,8-tetrakis(ethenyl)-2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane
Molecular Weight (g/mol) 344.66
PubChem CID 75706
Formula Weight 344.67
Percent Purity 97%
Refractive Index 1.434
Density 0.997
Chemical Name or Material 2,4,6,8-Tetramethyl-2,4,6,8-tetravinylcyclotetrasiloxane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'