missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trifluorobenzoyl chloride, 98%, Thermo Scientific™

Catalog Number p-7024255
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
88419-56-1
C7H2ClF3O
194.54
MFCD00061204
STBGCAUUOPNJBH-UHFFFAOYSA-N
2,4,5-trifluorobenzoylchloride, benzoyl chloride, 2,4,5-trifluoro, 2,4,5-tris fluoranyl benzoyl chloride, pubchem9716, acmc-209qt0, 2,4,5-trifluorobenzoyl-chloride, stbgcauuopnjbh-uhfffaoysa, timtec-bb sbb006657, 2,4,5-trifluoro-benzoyl chloride, 2,4,5-trifluorobenzoyl chloride
145164
2,4,5-trifluorobenzoyl chloride
FC1=CC(F)=C(C=C1F)C(Cl)=O
This item is not returnable. View return policy
88419-56-1
C7H2ClF3O
194.54
MFCD00061204
STBGCAUUOPNJBH-UHFFFAOYSA-N
2,4,5-trifluorobenzoylchloride, benzoyl chloride, 2,4,5-trifluoro, 2,4,5-tris fluoranyl benzoyl chloride, pubchem9716, acmc-209qt0, 2,4,5-trifluorobenzoyl-chloride, stbgcauuopnjbh-uhfffaoysa, timtec-bb sbb006657, 2,4,5-trifluoro-benzoyl chloride, 2,4,5-trifluorobenzoyl chloride
145164
2,4,5-trifluorobenzoyl chloride
FC1=CC(F)=C(C=C1F)C(Cl)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 1.52
Boiling Point 85°C (17 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.498
Quantity 1 g
UN Number UN3265
Beilstein 3606259
Sensitivity Moisture sensitive
Formula Weight 194.54
Percent Purity 98%
Chemical Name or Material 2,4,5-Trifluorobenzoyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'