missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloroaniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7025567
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
636-30-6
C6H4Cl3N
196.455
MFCD00007662
GUMCAKKKNKYFEB-UHFFFAOYSA-N
benzenamine, 2,4,5-trichloro, unii-7xkk8s8wo0, aniline, 2,4,5-trichloro, 7xkk8s8wo0, 2,4,5-trichlorophenylamine, ccris 2882, pubchem3213, 2,5-trichloroaniline, aniline,4,5-trichloro, 2,4,5-trichloro aniline
12487
2,4,5-trichloroaniline
C1=C(C(=CC(=C1Cl)Cl)Cl)N
This item is not returnable. View return policy
636-30-6
C6H4Cl3N
196.455
MFCD00007662
GUMCAKKKNKYFEB-UHFFFAOYSA-N
benzenamine, 2,4,5-trichloro, unii-7xkk8s8wo0, aniline, 2,4,5-trichloro, 7xkk8s8wo0, 2,4,5-trichlorophenylamine, ccris 2882, pubchem3213, 2,5-trichloroaniline, aniline,4,5-trichloro, 2,4,5-trichloro aniline
12487
2,4,5-trichloroaniline
C1=C(C(=CC(=C1Cl)Cl)Cl)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 92°C to 95°C
Boiling Point 269°C to 270°C
Quantity 50 g
Odor Characteristic
UN Number UN2018
Beilstein 879091
Formula Weight 196.46
Percent Purity 97%
Chemical Name or Material 2,4,5-Trichloroaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'