missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Fluorophenylamino)benzoic acid, 98%, Thermo Scientific™

Catalog Number p-7059044
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
54-59-1
C13H10FNO2
231.226
MFCD01675228
HYZIDPAMLUKNIR-UHFFFAOYSA-N
n-3-fluorophenyl anthranilic acid, 2-3-fluorophenyl amino benzoic acid, anthranilic acid, n-m-fluorophenyl, benzoic acid, 2-3-fluorophenyl amino, n-3-fluorophenyl anthranilsaeure german, acide n-3-flurophenyl anthranilique french, 2-3-fluorophenylamino benzoic acid, n-3-fluorophenyl anthranilsaeure, acide n-3-flurophenyl anthranilique
120158
2-(3-fluoroanilino)benzoic acid
C1=CC=C(C(=C1)C(=O)O)NC2=CC(=CC=C2)F
This item is not returnable. View return policy
54-59-1
C13H10FNO2
231.226
MFCD01675228
HYZIDPAMLUKNIR-UHFFFAOYSA-N
n-3-fluorophenyl anthranilic acid, 2-3-fluorophenyl amino benzoic acid, anthranilic acid, n-m-fluorophenyl, benzoic acid, 2-3-fluorophenyl amino, n-3-fluorophenyl anthranilsaeure german, acide n-3-flurophenyl anthranilique french, 2-3-fluorophenylamino benzoic acid, n-3-fluorophenyl anthranilsaeure, acide n-3-flurophenyl anthranilique
120158
2-(3-fluoroanilino)benzoic acid
C1=CC=C(C(=C1)C(=O)O)NC2=CC(=CC=C2)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 158°C to 163°C
Color Gray to Yellow
Quantity 2 g
Formula Weight 231.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2- (3-Fluorophenylamino)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'