Learn More
2,3-Dimethylphenylboronic acid, 97%, Thermo Scientific Chemicals
Catalog Number p-4228623
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
183158-34-1
C8H11BO2
149.99
MFCD01863524
ZYYANAWVBDFAHY-UHFFFAOYSA-N
2,3-dimethylbenzeneboronic acid, 2,3-dimethylphenyl boronic acid, o-xylene-3-boronic acid, 2,3-dimethylphenyl boranediol, 2,3-dimethylbenzeneboronicacid, 2,3-dimethylphenylboronicacid, pubchem5037, acmc-1bxk2, amtb394, ksc174k9l
2773395
(2,3-dimethylphenyl)boronic acid
B(C1=C(C(=CC=C1)C)C)(O)O
183158-34-1
C8H11BO2
149.99
MFCD01863524
ZYYANAWVBDFAHY-UHFFFAOYSA-N
2,3-dimethylbenzeneboronic acid, 2,3-dimethylphenyl boronic acid, o-xylene-3-boronic acid, 2,3-dimethylphenyl boranediol, 2,3-dimethylbenzeneboronicacid, 2,3-dimethylphenylboronicacid, pubchem5037, acmc-1bxk2, amtb394, ksc174k9l
2773395
(2,3-dimethylphenyl)boronic acid
B(C1=C(C(=CC=C1)C)C)(O)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 175.0°C
CAS Min % 96.0
Color White
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 149.99
Percent Purity 97%
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dimethylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2,3-Dimethylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.