missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dimethyl-1,3-butadiene, 98%, stab. with 100ppm BHT, Thermo Scientific Chemicals
466.82 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
2,3-Dimethyl-1,3-butadiene undergoes Diels Alder cycloaddition reaction and it also participates in polymerization reactions. It is also involved in manufacturing of synthetic rubber.
Solubility
Miscible with chloroform. Immiscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | -76°C |
Density | 0.726 |
Boiling Point | 68°C to 69°C |
Flash Point | −1°C (30°F) |
Refractive Index | 1.438 |
Quantity | 10 g |
UN Number | UN3295 |
Beilstein | 605285 |
Merck Index | 14,3238 |
Solubility Information | Miscible with chloroform. Immiscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'