Learn More
2,3-Dichloropyridine-4-boronic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used in HPLC and NMR chromatography. It is involved in several cross-coupling reactions.
Solubility
Soluble in organic solvents.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
It is used in HPLC and NMR chromatography. It is involved in several cross-coupling reactions.
Solubility
Soluble in organic solvents.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 136°C to 138°C |
Quantity | 250 mg |
Solubility Information | Soluble in organic solvents. |
Formula Weight | 191.81 |
Percent Purity | 95% |
Chemical Name or Material | 2,3-Dichloropyridine-4-boronic acid |