missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
10 g
Description
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Soluble in water. Slightly soluble in ethanol
Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.
It is employed as intermediate for pharmaceutical.
Solubility
Soluble in water. Slightly soluble in ethanol
Notes
Hygroscopic, store away from water/moisture. Keep the container tightly closed. Store away from oxidizing agent. Store under dry inert gas.
Specifications
Melting Point | 180°C to 190°C |
Color | White |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water; Slightly soluble in ethanol |
IUPAC Name | [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-[(2R,3R,4S,5R,6R)-4,5-diacetyloxy-2-(acetyloxymethyl)-6-bromooxan-3-yl]oxyoxan-2-yl]methyl acetate |
Formula Weight | 699.45 |
Physical Form | Powder |
Chemical Name or Material | 2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-cellobiosyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'