missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-beta-D-ribofuranose, 98%, Thermo Scientific Chemicals

Catalog Number p-7043209
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
89361-52-4
C26H28O5
420.505
MFCD01864312
NAQUAXSCBJPECG-VEYUFSJPSA-N
2,3,5-tri-o-benzyl-b-d-ribofuranose, 2,3,5-tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis phenylmethoxy-5-phenylmethoxymethyl, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
57369909
(2R,3R,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4
This item is not returnable. View return policy
89361-52-4
C26H28O5
420.505
MFCD01864312
NAQUAXSCBJPECG-VEYUFSJPSA-N
2,3,5-tri-o-benzyl-b-d-ribofuranose, 2,3,5-tri-o-benzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2-o,3-o,5-o-tribenzyl-beta-d-ribofuranose, 2r,3r,4r,5r-3,4-bis phenylmethoxy-5-phenylmethoxymethyl, 2r,3r,4r,5r-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol
57369909
(2R,3R,4R,5R)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
Solubility Information Soluble in chloroform
Optical Rotation +43° (c=2 in Dioxane)
Formula Weight 420.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-beta-D-ribofuranose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'