missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,3,4-Trifluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide, 95%

Brand:  Alfa Aesar™ H65443.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15429646

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

MFCD27975764
N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,3,4-trifluorobenzamide

Specifications

2,3,4-Trifluoro-N-(5,6,7,8-tetrahydro-1-naphthyl)benzamide
MFCD27975764
305.3
95%
1g
N-(5,6,7,8-Tetrahydro-1-naphthyl)-2,3,4-trifluorobenzamide
Solid
Videos
SDS Documents