missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorobenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-7081589
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1765-40-8
C7H2BrF5
260.989
MFCD00000299
XDEPVFFKOVDUNO-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
74484
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
C(C1=C(C(=C(C(=C1F)F)F)F)F)Br
This item is not returnable. View return policy
1765-40-8
C7H2BrF5
260.989
MFCD00000299
XDEPVFFKOVDUNO-UHFFFAOYSA-N
2,3,4,5,6-pentafluorobenzyl bromide, pentafluorobenzyl bromide, 1-bromomethyl-2,3,4,5,6-pentafluorobenzene, 1-bromomethylpentafluorobenzene, alpha-bromo-2,3,4,5,6-pentafluorotoluene, bromomethyl pentafluorobenzene, benzene, bromomethyl pentafluoro, 2,3,4,5,6-pentafluorobenzylbromide, pfbbr, pentafluorobenzylbromide
74484
1-(bromomethyl)-2,3,4,5,6-pentafluorobenzene
C(C1=C(C(=C(C(=C1F)F)F)F)F)Br

2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorobenzyl bromide is used as a reagent for derivatization of alcohols, carboxylic acids, sulfonamides for GC and polyfunctional thiols. It is used for the preparation of pentafluorobenzyl esters of organic acids for determination by capillary and GC. It is also used to derivatize N-7-substituted guanine adducts of DNA for determination by GC-electron capture-MS.

Solubility
Hydrolyzes in water.

Notes
Store in a cool, dry, well-ventilated area. Incompatible with bases and oxidizing agents. Store at 4°C. Protected from moisture.
TRUSTED_SUSTAINABILITY
Melting Point 18°C to 20°C
Density 1.844
Boiling Point 174°C to 175°C
Flash Point 83°C (181°F)
Refractive Index 1.472
Quantity 5 g
UN Number UN3265
Beilstein 647808
Solubility Information Hydrolyzes in water.
Formula Weight 261
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'