missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Ethoxyethoxy)ethyl acetate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acidity | 0.2 mg KOH/g max. |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -25.0°C |
Color | White to Beige |
Density | 1.0100g/mL |
Boiling Point | 210.0°C to 220.0°C |
Flash Point | 98°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Show More |
Safety and Handling
Product Identifier
- 2-(2-Ethoxyethoxy)ethyl acetate
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'