missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7020862
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
This item is not returnable. View return policy
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
3121
CHEBI:39867
2-propylpentanoic acid
CCCC(CCC)C(O)=O
This item is not returnable. View return policy
99-66-1
C8H16O2
144.21
MFCD00002672
NIJJYAXOARWZEE-UHFFFAOYSA-N
valproic acid, dipropylacetic acid, depakene, depakine, 2-propylvaleric acid, ergenyl, di-n-propylacetic acid, valproate, mylproin, pentanoic acid, 2-propyl
3121
CHEBI:39867
2-propylpentanoic acid
CCCC(CCC)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.905
Boiling Point 219°C to 220°C
Flash Point 126°C (258°F)
Odor Weak, Characteristic
Refractive Index 1.425
Quantity 10 g
UN Number UN3265
Beilstein 1750447
Merck Index 14,9913
Formula Weight 144.21
Percent Purity ≥98%
Chemical Name or Material 2,2-Di-n-propylacetic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'