Learn More
Alfa Aesar™ 2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile, 95%
Brand: Alfa Aesar™ H26022.03
Additional Details : CAS Number : 441060-95-3 Weight : 0.00100kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 1g |
---|
Chemical Identifiers
441060-95-3 | |
223.656 | |
PBMDCUAGVOXBMF-UHFFFAOYSA-N | |
22178154 | |
CC(=O)C(C#N)C1=C(C=C(C=C1)OC)Cl |
C11H10ClNO2 | |
MFCD08445711 | |
2-2-chloro-4-methoxyphenyl-3-oxobutanenitrile, 2-2-chloro-4-methoxyphenyl-3-oxobutyronitrile, acmc-1akfq, 1-cyano-1-2-chloro-4-methoxyphenyl propan-2-on | |
2-(2-chloro-4-methoxyphenyl)-3-oxobutanenitrile |
Specifications
2-(2-Chloro-4-methoxyphenyl)-3-oxobutyronitrile | |
1g | |
C11H10ClNO2 | |
UN3439 | |
PBMDCUAGVOXBMF-UHFFFAOYSA-N | |
2-(2-chloro-4-methoxyphenyl)-3-oxobutanenitrile | |
22178154 | |
95% |
78°C to 80°C | |
441060-95-3 | |
MFCD08445711 | |
2-2-chloro-4-methoxyphenyl-3-oxobutanenitrile, 2-2-chloro-4-methoxyphenyl-3-oxobutyronitrile, acmc-1akfq, 1-cyano-1-2-chloro-4-methoxyphenyl propan-2-on | |
CC(=O)C(C#N)C1=C(C=C(C=C1)OC)Cl | |
223.656 | |
223.66 |