Learn More
2-(2-Bromoethoxy)tetrahydro-2H-pyran, 96%, stabilized, Thermo Scientific Chemicals
Catalog Number p-3771021
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
17739-45-6
C7H13BrO2
209.08
MFCD01321310
GCUOLJOTJRUDIZ-UHFFFAOYSA-N
2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran
86621
2-(2-bromoethoxy)oxane
C1CCOC(C1)OCCBr
17739-45-6
C7H13BrO2
209.08
MFCD01321310
GCUOLJOTJRUDIZ-UHFFFAOYSA-N
2-2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy oxane, 2h-pyran, 2-2-bromoethoxy tetrahydro, 2-2-bromo-ethoxy-tetrahydro-pyran, 2-2-bromoethoxy tetrahydropyran, 2 2-bromoethoxy tetrahydro-2h-pyran, 2-2-bromoethoxy-tetrahydro-2h-pyran, 2h-pyran,3-2-bromoethoxy tetrahydro, 2-2-bromoethyl oxy tetrahaydro-2h-pyran
86621
2-(2-bromoethoxy)oxane
C1CCOC(C1)OCCBr
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Density 1.3840g/mL
Boiling Point 62°C to 64°C (0.4 mmHg)
Flash Point 87°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 1.384
Solubility Information Solubility in water: insoluble
Formula Weight 209.08
Percent Purity 96%
Chemical Name or Material 2-(2-Bromoethoxy)tetrahydro-2H-pyran
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Bromoethoxy)tetrahydro-2H-pyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.