missing translation for 'onlineSavingsMsg'
Learn More

2,2,6,6-Tetramethyl-3,5-heptanedione, 98%, Thermo Scientific Chemicals

Catalog Number p-7022373
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1118-71-4
C11H20O2
184.279
MFCD00008848
YRAJNWYBUCUFBD-UHFFFAOYSA-N
dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497
70700
2,2,6,6-tetramethylheptane-3,5-dione
CC(C)(C)C(=O)CC(=O)C(C)(C)C
This item is not returnable. View return policy
1118-71-4
C11H20O2
184.279
MFCD00008848
YRAJNWYBUCUFBD-UHFFFAOYSA-N
dipivaloylmethane, 2,2,6,6-tetramethyl-3,5-heptanedione, 3,5-heptanedione, 2,2,6,6-tetramethyl, unii-r8ui909hoy, tmhd, 2,2,6,6-tetramethyl-3,5-heptanedione dipivaloylmethane, r8ui909hoy, 2,2,6,6-tetramethyl-heptane-3,5-dione, pubchem12497
70700
2,2,6,6-tetramethylheptane-3,5-dione
CC(C)(C)C(=O)CC(=O)C(C)(C)C

2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,6,6-Tetramethyl-3,5-heptanedione is used in the synthesis of α-aryl-β-diketones1 and dicyanamidobenzene-bridge diruthenium complex.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 0.889
Boiling Point 72°C to 73°C (6 mmHg)
Flash Point 67°C (152°F)
Refractive Index 1.459
Quantity 5 g
Beilstein 1447269
Solubility Information Difficult to mix.
Formula Weight 184.28
Percent Purity 98%
Chemical Name or Material 2,2,6,6-Tetramethyl-3,5-heptanedione
compliance-icons
Product Identifier
  • 2,2,6,6-Tetramethyl-3,5-heptanedione
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'