missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,2-Trimethylacetophenone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solubility
Not miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
Not miscible with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
Specifications
Density | 0.97 |
Boiling Point | 219°C to 222°C |
Flash Point | 87°C (188°F) |
Refractive Index | 1.5084 |
Quantity | 1 g |
Beilstein | 1906460 |
Solubility Information | Not miscible with water. |
Formula Weight | 162.23 |
Percent Purity | 98% |
Chemical Name or Material | 2,2,2-Trimethylacetophenone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'