Learn More
2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific Chemicals
Description
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,2,2-Trichloroethyl chloroformate is used as a protecting reagent for aliphatic and aromatic hydroxyl and amino groups. It is used as derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples, during N-demethylation of dextromethorphan. It was used as starting reagent during the synthesis of 6-Nor-9,10-dihydrolysergic acid methyl ester.
Solubility
Soluble in Ether, Benzene, Chloroform, Ethanol. Insoluble in water.
Notes
Moisture Sensitive. Desiccate at 4°C. Store under inert gas. Incompatible materials are Strong oxidizing agents.
Specifications
Melting Point | 0°C to 2°C |
Density | 1.539 |
Boiling Point | 171°C to 172°C |
Flash Point | >100°C (212°F) |
Refractive Index | 1.47 |
Quantity | 25 g |
UN Number | UN3277 |
Beilstein | 970619 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in Ether,Benzene,Chloroform,Ethanol. Insoluble in water. |
Show More |
RUO – Research Use Only