missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trichloroethyl chloroformate, 97%, Thermo Scientific Chemicals

Catalog Number p-203086
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17341-93-4
MFCD00000810
LJCZNYWLQZZIOS-UHFFFAOYSA-N
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
87063
2,2,2-trichloroethyl carbonochloridate
C(C(Cl)(Cl)Cl)OC(=O)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
17341-93-4
MFCD00000810
LJCZNYWLQZZIOS-UHFFFAOYSA-N
2,2,2-trichloroethyl chloroformate, trichloroethyl chloroformate, 2,2,2-trichloroethylchloroformate, carbonochloridic acid, 2,2,2-trichloroethyl ester, 2,2,2-trichloroethoxycarbonyl chloride, chloroformic acid 2,2,2-trichloroethyl ester, formic acid, chloro-, 2,2,2-trichloroethyl ester, chloro 2,2,2-trichloroethoxy methanone, beta, beta, beta-trichloroethoxycarbonyl chloride, troc
87063
2,2,2-trichloroethyl carbonochloridate
C(C(Cl)(Cl)Cl)OC(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 1°C
Color Yellow
Density 1.5390g/mL
Boiling Point 171°C to 172°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H2Cl4O2
Linear Formula ClCO2CH2CCl3
Refractive Index 1.4690 to 1.471
Quantity 25 g
Fieser 02,426; 03,296; 04,522; 05,686; 07,383
Specific Gravity 1.539
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in most common organic solvents
Molecular Weight (g/mol) 211.86
Formula Weight 211.86
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trichloroethyl chloroformate, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trichloroethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P402+P404-Store in a dry place. Store in a closed container.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'