missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-indol-3-yl)ethan-1-ol, Thermo Scientific™

Catalog Number p-4391373
Click to view available options
Quantity:
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
526-55-6
C10H11NO
161.20
MFCD00005659
MBBOMCVGYCRMEA-UHFFFAOYSA-N
tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
10685
CHEBI:17890
2-(1H-indol-3-yl)ethanol
OCCC1=CNC2=CC=CC=C12
This item is not returnable. View return policy
526-55-6
C10H11NO
161.20
MFCD00005659
MBBOMCVGYCRMEA-UHFFFAOYSA-N
tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
10685
CHEBI:17890
2-(1H-indol-3-yl)ethanol
OCCC1=CNC2=CC=CC=C12
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Merck Index 15, 9978
Formula Weight 161.2
Percent Purity 97%
Chemical Name or Material 2-(1H-Indol-3-yl)ethan-1-ol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'