missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Cyclohexenyl)cyclohexanone, 85+%, cont. ca 10% 2-cyclohexylidenecyclohexanone, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
It is a perfuming agent. It is used for special solvents and as a raw material in agrochemicals and dyestuffs.
Solubility
Soluble in water. Insoluble in alcohol.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Density | 0.998 |
Boiling Point | 265°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.506 |
Quantity | 25 g |
Beilstein | 1308501 |
Solubility Information | Soluble in water. Insoluble in alcohol. |
IUPAC Name | 2-(cyclohexen-1-yl)cyclohexan-1-one |
Formula Weight | 178.28 |
Percent Purity | ≥85% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'