Learn More
(1R,3r,5S)-8-Azabicyclo[3.2.1]oct-3-yl Hydroxydiphenylacetate Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(1R,3r,5S)-8-Azabicyclo[3.2.1]oct-3-yl Hydroxydiphenylacetate Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (1R,3r,5S)-8-Azabicyclo(3.2.1)oct-3-yl Hydroxydiphenylacetate Hydrochloride |
| CAS | 63516-30-3 |
| Analyte or Component Names | (1R,3r,5S)-8-Azabicyclo(3.2.1)oct-3-yl Hydroxydiphenylacetate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H23 N O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Trospium Chloride Imp. B (EP), Trospium Chloride USP Related Compound B, Trospium Chloride USP RC B, Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, (3-endo)-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, (3-endo)-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (9CI), Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, 8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride, endo-, Trospium Related Compound B as Hydrochloride, Trospium Chloride Impurity B as Hydrochloride, Trospium Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C21H23NO3.ClH/c23-20(25-19-13-17-11-12-18(14-19)22-17)21(24,15-7-3-1-4-8-15)16-9-5-2-6-10-16;/h1-10,17-19,22,24H,11-14H2;1H/t17-,18+,19+; |
| SMILES | Cl.OC(C(=O)O[C@H]1C[C@H]2CC[C@@H](C1)N2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | [(1S,5R)-8-azabicyclo[3.2.1]octan-3-yl] 2-hydroxy-2,2-diphenylacetate;hydrochloride |
| Show More |