Learn More
Alfa Aesar™ 1H,1H,2H,2H-Perfluorodecylmethyldichlorosilane, 95%
Brand: Alfa Aesar™ L16583.14
Additional Details : CAS Number : 3102-79-2 Weight : 0.02500kg Transport : UN number : 2987 Chem class : 8 Pack group : II
Quantity | 25g |
---|
Chemical Identifiers
3102-79-2 | |
561.135 | |
PVBMWIXRKLGXPI-UHFFFAOYSA-N | |
137826 | |
C[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(Cl)Cl |
C11H7Cl2F17Si | |
MFCD00042342 | |
1h,1h,2h,2h-perfluorodecylmethyldichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl methyldichlorosilane, dichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl methylsilane, dichloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-methylsilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-methyldichlorosilane, 1h,1h,2h,2h-perfluorodecyldichloromethylsilane, bis chloranyl-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecakis fluoranyl decyl-methyl-silane | |
dichloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-methylsilane |
Specifications
1H,1H,2H,2H-Perfluorodecylmethyldichlorosilane | |
C11H7Cl2F17Si | |
UN2987 | |
1h,1h,2h,2h-perfluorodecylmethyldichlorosilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl methyldichlorosilane, dichloro 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl methylsilane, dichloro-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl-methylsilane, heptadecafluoro-1,1,2,2-tetrahydrodecyl-1-methyldichlorosilane, 1h,1h,2h,2h-perfluorodecyldichloromethylsilane, bis chloranyl-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecakis fluoranyl decyl-methyl-silane | |
C[Si](CCC(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(Cl)Cl | |
561.135 | |
561.14 | |
1.63 | |
Moisture sensitive | |
25g |
3102-79-2 | |
MFCD00042342 | |
5889048 | |
PVBMWIXRKLGXPI-UHFFFAOYSA-N | |
dichloro-(3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecyl)-methylsilane | |
137826 | |
95% | |
1.346 | |
205°C to 207°C |