Learn More
16alpha,17-[1-Methylethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione (Desonide), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
16alpha,17-[1-Methylethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione (Desonide)
Specifications
Specifications
| Chemical Name or Material | Desonide |
| CAS | 638-94-8 |
| Analyte or Component Names | Desonide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H32 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Budesonide Imp. F (EP), Desonide, 16alpha,17-[1-Methylethylidenebis(oxy)]-11beta,21-dihydroxypregna-1,4-diene-3,20-dione, Budesonide Impurity F |
| InChI Formula | InChI=1S/C24H32O6/c1-21(2)29-19-10-16-15-6-5-13-9-14(26)7-8-22(13,3)20(15)17(27)11-23(16,4)24(19,30-21)18(28)12-25/h7-9,15-17,19-20,25,27H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,20+,22-,23-,24+/m0/s1 |
| SMILES | CC1(C)O[C@@H]2C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO |
| IUPAC Name | 16α,17-[1-Methylethylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione |
| Show More |