missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 11-(Pentafluorophenoxy)undecyltrimethoxysilane, 95%

1438.29 NOK - 3996.76 NOK

Chemical Identifiers

CAS 944721-47-5
Molecular Formula C20H31F5O4Si
Molecular Weight (g/mol) 458.541
MDL Number MFCD13181913
InChI Key OPIXAGPRZGMNHK-UHFFFAOYSA-N
Synonym 11-pentafluorophenoxyundecyl trimethoxysilane, 11-pentafluorophenoxy undecyltrimethoxysilane, trimethoxy 11-perfluorophenoxy undecyl silane, 11-pentafluorophenoxy undecyl trimethoxysilane, trimethoxy 11-2,3,4,5,6-pentafluorophenoxy undecyl silane
PubChem CID 23582154
IUPAC Name trimethoxy-[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane
SMILES CO[Si](CCCCCCCCCCCOC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15488595 SDS View Documents Alfa Aesar™
H33702.MD
250mg 1438.29 NOK
250mg View alternative products
Save Up to 
»
Estimated Shipment
22-07-2020
Add to basket
 
15478595 SDS View Documents Alfa Aesar™
H33702.03
1g 3996.76 NOK
1g View alternative products
Save Up to 
»
Estimated Shipment
22-07-2020
Add to basket
 
Specifications

Chemical Identifiers

944721-47-5
458.541
OPIXAGPRZGMNHK-UHFFFAOYSA-N
23582154
CO[Si](CCCCCCCCCCCOC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC
C20H31F5O4Si
MFCD13181913
11-pentafluorophenoxyundecyl trimethoxysilane, 11-pentafluorophenoxy undecyltrimethoxysilane, trimethoxy 11-perfluorophenoxy undecyl silane, 11-pentafluorophenoxy undecyl trimethoxysilane, trimethoxy 11-2,3,4,5,6-pentafluorophenoxy undecyl silane
trimethoxy-[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane

Specifications

11-(Pentafluorophenoxy)undecyltrimethoxysilane
C20H31F5O4Si
11-pentafluorophenoxyundecyl trimethoxysilane, 11-pentafluorophenoxy undecyltrimethoxysilane, trimethoxy 11-perfluorophenoxy undecyl silane, 11-pentafluorophenoxy undecyl trimethoxysilane, trimethoxy 11-2,3,4,5,6-pentafluorophenoxy undecyl silane
CO[Si](CCCCCCCCCCCOC1=C(C(=C(C(=C1F)F)F)F)F)(OC)OC
458.541
458.54
1.4370-1.4410
130°C to 135°C (0.04mmHg)
944721-47-5
MFCD13181913
OPIXAGPRZGMNHK-UHFFFAOYSA-N
trimethoxy-[11-(2,3,4,5,6-pentafluorophenoxy)undecyl]silane
23582154
95%
Moisture sensitive
250mg