missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-Hydroxybenzo[h]quinoline, 98%, Alfa Aesar™
Click to view available options
Quantity:
5 g
25 g
Description
Applications
10-Hydroxybenzo[h]quinolines is used to detect boronic acids in reactions and boron containing compounds on solid support. It is also used as a proton transfer-fluorescence probe to be applied to human serum albumin and beaver apomyoglobin.
Solubility
Soluble in acetone, chloroform and ethyl acetate.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
10-Hydroxybenzo[h]quinolines is used to detect boronic acids in reactions and boron containing compounds on solid support. It is also used as a proton transfer-fluorescence probe to be applied to human serum albumin and beaver apomyoglobin.
Solubility
Soluble in acetone, chloroform and ethyl acetate.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 103°C to 105°C |
Quantity | 5 g |
Beilstein | 141770 |
Solubility Information | Soluble in acetone,chloroform and ethyl acetate. |
Formula Weight | 195.22 |
Percent Purity | 98% |
Chemical Name or Material | 10-Hydroxybenzo[h]quinoline |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'