Learn More
1-(Trimethylsilyl)-1H-benzotriazole, 97%, Thermo Scientific™
Catalog Number p-4228942
Click to view available options
Quantity:
1 g
5 g
43183-36-4
C9H13N3Si
191.31
VHOLOZYRMQVHBK-UHFFFAOYSA-N
1-trimethylsilyl-1h-benzotriazole, 1-trimethylsilylbenzotriazole, 1-trimethylsilyl-1h-benzo d 1,2,3 triazole, 1-trimethylsilyl-1,2,3-benzotriazole, 1-tms-1h-benzotriazole, acmc-1bn8x, n-trimethylsilylbenzotriazole, benzotriazol-1-yl trimethyl silane, 1-trimethylsilanyl-1h-benzotriazole
3558326
benzotriazol-1-yl(trimethyl)silane
C[Si](C)(C)N1C2=CC=CC=C2N=N1
43183-36-4
C9H13N3Si
191.31
VHOLOZYRMQVHBK-UHFFFAOYSA-N
1-trimethylsilyl-1h-benzotriazole, 1-trimethylsilylbenzotriazole, 1-trimethylsilyl-1h-benzo d 1,2,3 triazole, 1-trimethylsilyl-1,2,3-benzotriazole, 1-tms-1h-benzotriazole, acmc-1bn8x, n-trimethylsilylbenzotriazole, benzotriazol-1-yl trimethyl silane, 1-trimethylsilanyl-1h-benzotriazole
3558326
benzotriazol-1-yl(trimethyl)silane
C[Si](C)(C)N1C2=CC=CC=C2N=N1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0540g/mL
Boiling Point 125.0°C to 126.0°C (3.0 mmHg)
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.054
Formula Weight 191.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(Trimethylsilyl)-1H-benzotriazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(Trimethylsilyl)-1H-benzotriazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.