missing translation for 'onlineSavingsMsg'
Learn More

(1-Tetradecyl)trimethylammonium chloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7045920
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
4574-04-3
MFCD00059968
This item is not returnable. View return policy
4574-04-3
MFCD00059968

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Molecular Formula C17H38ClN
Quantity 5 g
Beilstein 3916572
Sensitivity Hygroscopic
Synonym tetradecyltrimethylammonium chloride, myristyltrimethylammonium chloride, n,n,n-trimethyltetradecan-1-aminium chloride, trimethyltetradecylammonium chloride, 1-tetradecanaminium, n,n,n-trimethyl-, chloride, n-tetradecyltrimethylammonium chloride, rmv7nju3fx, unii-rmv7nju3fx, trimethyl tetradecyl azanium chloride, tetradecyl trimethyl ammonium chloride
InChI Key CEYYIKYYFSTQRU-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCC[N+](C)(C)C.[Cl-]
IUPAC Name trimethyl(tetradecyl)azanium;chloride
Molecular Weight (g/mol) 291.95
PubChem CID 20708
Formula Weight 291.95
Percent Purity 98%
Chemical Name or Material (1-Tetradecyl)trimethylammonium chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'